Learn More
MSC Concrobium Mold Control, Liquid, Ready to Use, EPA-Registered, 1 gal.
Catalog No. 502817458
Encompass
Concrobium Mold Control is an odorless, colorless liquid solution designed to effectively eliminate and prevent mold and musty odors. This EPA-registered formula works by crushing mold spores at their roots as it dries, leaving an invisible antimicrobial shield to inhibit future mold growth without harmful chemicals like bleach, ammonia, or volatile organic compounds (VOCs).
  • Eliminates, cleans, and prevents mold and musty odors
  • Contains no bleach, ammonia, or VOCs
  • Penetrates to the roots of mold
  • EPA-registered for effective mold control
  • Works on various surfaces including drywall, concrete, wood, and tile
  • Ready to use; no dilution required
  • Maintains protection against re-growth without rinsing
Catalog No. 50-281-7458 Supplier MSC Supplier No. 12168258
Concrobium Mold Control is an odorless, colorless liquid solution designed to effectively eliminate and prevent mold and musty odors. This EPA-registered formula works by crushing mold spores at their roots as it dries, leaving an invisible antimicrobial shield to inhibit future mold growth without harmful chemicals like bleach, ammonia, or volatile organic compounds (VOCs).
  • Eliminates, cleans, and prevents mold and musty odors
  • Contains no bleach, ammonia, or VOCs
  • Penetrates to the roots of mold
  • EPA-registered for effective mold control
  • Works on various surfaces including drywall, concrete, wood, and tile
  • Ready to use; no dilution required
  • Maintains protection against re-growth without rinsing