Learn More
Electron Microscopy Sciences Cold Mounting Disposable Mold Cups, 1.25"
SDP
Catalog No. 50997419
Encompass_Preferred
Mounting cups for cold mounting/embedding. Polyethylene, one part molds. Closed one end. Red.
Size: 1.25"
Catalog No. 50-997-419 Supplier Electron Microscopy Sciences Supplier No. 5048015
Mounting cups for cold mounting/embedding. Polyethylene, one part molds. Closed one end. Red.
Size: 1.25"