Learn More
Electron Microscopy Sciences Cold Mounting Disposable Mold Cups, 1.5"
SDP
Catalog No. 50997420
Encompass_Preferred
Mounting cups for cold mounting/embedding. Polyethylene, one part molds. Closed one end. Red.
Size: 1½"
Catalog No. 50-997-420 Supplier Electron Microscopy Sciences Supplier No. 5048020
Mounting cups for cold mounting/embedding. Polyethylene, one part molds. Closed one end. Red.
Size: 1½"