Learn More
Electron Microscopy Sciences EMS Disposable Multi-Well Embedding Mold
SDP
Catalog No. 501927461
Encompass_Preferred
A clear flexible LDPE mold. A great idea if you do not want to clean the molds. Same as our flat embedding silicone molds. 2 Troughs: 1"(L) x 1/8"(W) x 1/8"(D) 14 Cavities: 1/4"(L) x 1/2"(W) x 1/8"(D) 1 Trough: 2 3/4"(L) x 3/32"(W) x 1/8"(D) 10 Cavities: 3/8"(L) x 3/4"(W) x 1/8"(D)
Catalog No. 50-192-7461 Supplier Electron Microscopy Sciences Supplier No. 70919
A clear flexible LDPE mold. A great idea if you do not want to clean the molds. Same as our flat embedding silicone molds. 2 Troughs: 1"(L) x 1/8"(W) x 1/8"(D) 14 Cavities: 1/4"(L) x 1/2"(W) x 1/8"(D) 1 Trough: 2 3/4"(L) x 3/32"(W) x 1/8"(D) 10 Cavities: 3/8"(L) x 3/4"(W) x 1/8"(D)