Learn More
Electron Microscopy Sciences Mold for Long Tissues
SDP
Catalog No. 5029272
Encompass_Preferred
Accommodates longer pieces of tissues and makes it easy to embed long specimens and orient them properly. Tissues lie flat in the well. Ideal for any specimen that needs immediate sectioning (e.g. cell monolayer), 24 cavities
Catalog No. 50-292-72 Supplier Electron Microscopy Sciences Supplier No. 70166
Accommodates longer pieces of tissues and makes it easy to embed long specimens and orient them properly. Tissues lie flat in the well. Ideal for any specimen that needs immediate sectioning (e.g. cell monolayer), 24 cavities