Learn More
Polysciences, Inc. Peel-A-WayEmbedding Mold (Square - S22)
SDP
Catalog No. 50465347
Encompass
Peel-a-Waya Embedding Mold (Square - S22). Disposable paraffin molds designed to in.Peel-a-Way in. from the solidified embedded block. Be sure to consider Polyciences' Polyfin and Peel-a-Way Micro-Cut Paraffin for use with Peel-a-Way molds. May also be used for epoxy and acrylic resin embedding procedures. Features: - Polyethylene - 22mm x 22mm square - 20mm deep - 288 molds per case
Catalog No. 50-465-347 Supplier Polysciences, Inc. Supplier No. 18646A1
Peel-a-Waya Embedding Mold (Square - S22). Disposable paraffin molds designed to in.Peel-a-Way in. from the solidified embedded block. Be sure to consider Polyciences' Polyfin and Peel-a-Way Micro-Cut Paraffin for use with Peel-a-Way molds. May also be used for epoxy and acrylic resin embedding procedures. Features: - Polyethylene - 22mm x 22mm square - 20mm deep - 288 molds per case