Learn More
Polysciences, Inc. Peel-A-Way Embedding Mold (Rectangular - R30), 288/cs
SDP
Catalog No. 50465346
Encompass
Disposable paraffin molds designed to "Peel-A-Way" from the solidified embedded block. Polysciences Peel-A-Way Embedding Molds are useful for plastic, frozen, and epoxy embedding. The unique polyethylene construction is designed to Peel-A-Way from the solidified block, eliminating the need for release agents and block trimming.
Catalog No. 50-465-346 Supplier Polysciences, Inc. Supplier No. 18646B1
Disposable paraffin molds designed to "Peel-A-Way" from the solidified embedded block. Polysciences Peel-A-Way Embedding Molds are useful for plastic, frozen, and epoxy embedding. The unique polyethylene construction is designed to Peel-A-Way from the solidified block, eliminating the need for release agents and block trimming.