Learn More
Polysciences, Inc. Mold, Peel-A-Way Embedding; Truncated - T8; Size:22 x 22 sq. mm truncated to 8 x 8mm; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip
SDP
Catalog No. NC9991740
Encompass_Preferred
Mold, Peel-A-Way Embedding; Truncated - T8; Size:22 x 22 sq. mm truncated to 8 x 8mm; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip
Catalog No. NC9991740 Supplier Polysciences, Inc. Supplier No. 189851
Mold, Peel-A-Way Embedding; Truncated - T8; Size:22 x 22 sq. mm truncated to 8 x 8mm; Clear plastic weighing tray; Embedding paraffin; Disposable embedding mold; Tissue capsule; Slide grip