Learn More
Electron Microscopy Sciences Individual Polyethylene Molds, 16 x 8 mm
SDP
Catalog No. 5034088
Encompass_Preferred
Molded from polyethylene, three popular sizes are available to meet your needs. These Individual Molds can be used with EBH2 Block Holders, Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Suitable for JB-4, JB-4-Plus, Immuno-Bed, LR White, LR Gold or Paraffin embedding.
Size: 16 x 8 mm
Catalog No. 50-340-88 Supplier Electron Microscopy Sciences Supplier No. 7017716
Molded from polyethylene, three popular sizes are available to meet your needs. These Individual Molds can be used with EBH2 Block Holders, Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Suitable for JB-4, JB-4-Plus, Immuno-Bed, LR White, LR Gold or Paraffin embedding.
Size: 16 x 8 mm