Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC PTFE Dry-Film Lubricant, Min of 4
SDP
Supplier:  MILLER STEPHENSON CHEMICAL COMPANY INC MS122XD
Encompass_Preferred
MS-122XD was designed as an efficient, durable, and universal dry film lubricant with superior release and non-stick characteristics. Has the fastest dry time of all the products in the 122 series with the most refined final ultra-thin film properties.
Catalog No. NC9984747