Learn More
Polysciences, Inc. Peel-a-Way Embedding Mold (Truncated - T12). Disposable paraffin molds designed to in.Peel-a-Way in. from the solidified embedded block.
SDP
Catalog No. 50465348
Encompass
Peel-a-Way Embedding Mold (Truncated - T12). Disposable paraffin molds designed to in.Peel-a-Way in. from the solidified embedded block. Be sure to consider Polyciences' Polyfin and Peel-a-Way Micro-Cut Paraffin for use with Peel-a-Way molds. May also be used for epoxy and acrylic resin embedding procedures. Polyethylene 22mm x 22mm square, truncated to 12mm x 12mm 20mm deep 288 molds per case
Catalog No. 50-465-348 Supplier Polysciences, Inc. Supplier No. 189861
Peel-a-Way Embedding Mold (Truncated - T12). Disposable paraffin molds designed to in.Peel-a-Way in. from the solidified embedded block. Be sure to consider Polyciences' Polyfin and Peel-a-Way Micro-Cut Paraffin for use with Peel-a-Way molds. May also be used for epoxy and acrylic resin embedding procedures. Polyethylene 22mm x 22mm square, truncated to 12mm x 12mm 20mm deep 288 molds per case