Learn More
Biophysics Assay Laboratory Inc MOLDAY ION RHODAMINE B MIRB
SDP
Supplier:  Biophysics Assay Laboratory Inc CL50Q016A50
Encompass
This is a 35 nm rhodamine B-labeled iron-based superparamagnetic contrast agent (USPIO) packaged in a 2 ml sealed serum bottle containing 0.5 ml or 2 ml of the product. Each bottle has a 2 mg Fe/ml concentration having a zeta potential of ~ +31 mV.
Catalog No. NC1924688