Learn More
Electron Microscopy Sciences Molded, 100- Microscope Slide Box
SDP
Supplier:  Electron Microscopy Sciences 71473
Encompass_Preferred
Molded one-piece polypropylene construction with an integral living hinge. Numbering in the base corresponds to the printed index in the covers for slide identification purpose. Boxes are designed to be stackable to save space. The tensioned clasp insures a secure closure. Fits 1 x 3" microscope slides.
Catalog No. 50-931-116