Learn More
Grainger Molded Grating Span 10 ft.
Catalog No. 502767581
Encompass_Preferred
The Grainger 16U512 is a 5-ton capacity hydraulic bottle jack made of heavy duty steel with a wide base for stability. It has a built-in overload valve to prevent over-extension and can lift loads up to 10 inches high. This hydraulic jack has a handle that allows the user to pump and raise loads easily. It meets ASME PALD standards and has a one year warranty from Grainger. The jack weighs 14 pounds and has a maximum height of 17 inches.
Catalog No. 50-276-7581 Supplier Grainger Supplier No. 16U512
The Grainger 16U512 is a 5-ton capacity hydraulic bottle jack made of heavy duty steel with a wide base for stability. It has a built-in overload valve to prevent over-extension and can lift loads up to 10 inches high. This hydraulic jack has a handle that allows the user to pump and raise loads easily. It meets ASME PALD standards and has a one year warranty from Grainger. The jack weighs 14 pounds and has a maximum height of 17 inches.