Learn More
Grainger Molded Grating Span 10 ft.
Catalog No. 502768360
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester resin. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a load depth of 1-1/2". The grating has a maximum uniform load of 100 psf and a concentrated load of 300 lbs. It meets OSHA and ADA requirements for slip resistance. The grating panels are 2' wide and up to 24' long. This strong, lightweight FRP grating is suitable for industrial platforms, walkways, stairs and more.
Catalog No. 50-276-8360 Supplier Grainger Supplier No. 16U491
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester resin. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a load depth of 1-1/2". The grating has a maximum uniform load of 100 psf and a concentrated load of 300 lbs. It meets OSHA and ADA requirements for slip resistance. The grating panels are 2' wide and up to 24' long. This strong, lightweight FRP grating is suitable for industrial platforms, walkways, stairs and more.