Learn More
Grainger Molded Grating Span 10 ft.
Catalog No. 502764883
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester resin. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a thickness of 1-1/4". The grating has a maximum load capacity of 100 psf and meets OSHA standards for slip resistance. It can be used for flooring, walkways, platforms, and more. The grating comes in 4' x 8' sheets and can be fabricated for custom sizes and configurations.
Catalog No. 50-276-4883 Supplier Grainger Supplier No. 16U493
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester resin. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a thickness of 1-1/4". The grating has a maximum load capacity of 100 psf and meets OSHA standards for slip resistance. It can be used for flooring, walkways, platforms, and more. The grating comes in 4' x 8' sheets and can be fabricated for custom sizes and configurations.