Learn More
Grainger Molded Grating Span 10 ft.
Catalog No. 502766248
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester or vinyl ester resins. It has a standard mesh size of 1" x 3/16" and comes in a gray color. The grating has a load capacity of 100 psf with a maximum deflection of 1/4" and flame spread rating of Class A. It is designed for use in corrosive environments and has low electrical and thermal conductivity. The standard panel size is 4' x 8' and it can be fabricated to custom sizes. It has a light weight of 3 lbs/ft2 which allows for easy installation.
Catalog No. 50-276-6248 Supplier Grainger Supplier No. 16U514
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester or vinyl ester resins. It has a standard mesh size of 1" x 3/16" and comes in a gray color. The grating has a load capacity of 100 psf with a maximum deflection of 1/4" and flame spread rating of Class A. It is designed for use in corrosive environments and has low electrical and thermal conductivity. The standard panel size is 4' x 8' and it can be fabricated to custom sizes. It has a light weight of 3 lbs/ft2 which allows for easy installation.