Learn More
Grainger Molded Grating Span 12 ft.
Catalog No. 502765624
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength fiberglass reinforced plastic (FRP) molded grating. It is made of a matrix of continuous rovings of glass fibers bonded with a thermosetting resin. It has a load capacity of 100 psf with a maximum span of 4 feet. The grating has a matte gray finish and is available in a standard mesh size of 1 inch. It can be used for flooring, walkways, platforms, stair treads and other applications requiring an anti-slip, corrosion resistant surface. The molded grating panels measure 4x8 feet and weigh approximately 8 lbs per square foot.
Catalog No. 50-276-5624 Supplier Grainger Supplier No. 16U502
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength fiberglass reinforced plastic (FRP) molded grating. It is made of a matrix of continuous rovings of glass fibers bonded with a thermosetting resin. It has a load capacity of 100 psf with a maximum span of 4 feet. The grating has a matte gray finish and is available in a standard mesh size of 1 inch. It can be used for flooring, walkways, platforms, stair treads and other applications requiring an anti-slip, corrosion resistant surface. The molded grating panels measure 4x8 feet and weigh approximately 8 lbs per square foot.