Learn More
Grainger Molded Grating Span 12 ft.
Catalog No. 502768322
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/8" openings. This grating is designed for use in corrosive environments and offers an alternative to traditional metallic gratings. With its lightweight design, the Fibergrate grating is easy to fabricate, install and maintain. It can be used for flooring, walkways, platforms, stairs and more. This molded grating measures 4' x 8' and has a load capacity of 100 psf with a maximum deflection of 1/4"
Catalog No. 50-276-8322 Supplier Grainger Supplier No. 16U499
Add to Cart
Add to Cart
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/8" openings. This grating is designed for use in corrosive environments and offers an alternative to traditional metallic gratings. With its lightweight design, the Fibergrate grating is easy to fabricate, install and maintain. It can be used for flooring, walkways, platforms, stairs and more. This molded grating measures 4' x 8' and has a load capacity of 100 psf with a maximum deflection of 1/4"