Learn More
Grainger Molded Grating Span 3 ft PK2
Catalog No. 502766544
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength fiberglass reinforced plastic grating made by pultrusion. It has a matte gray finish and is available in a variety of standard sizes and mesh configurations. The grating has a flame spread rating of 25 or less per ASTM E-84 and is designed for use in corrosive environments. It provides an anti-slip walking surface.
Catalog No. 50-276-6544 Supplier Grainger Supplier No. 16U507
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength fiberglass reinforced plastic grating made by pultrusion. It has a matte gray finish and is available in a variety of standard sizes and mesh configurations. The grating has a flame spread rating of 25 or less per ASTM E-84 and is designed for use in corrosive environments. It provides an anti-slip walking surface.