Learn More
Grainger Molded Grating Span 3 ft PK2
Catalog No. 502767014
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/16" bearing bars at 1-3/16" centers. The panels measure 4' x 8' and have a load capacity of 100 psf. This grating is designed for use in corrosive environments and provides an alternative to traditional metallic gratings. The molded grating has low thermal and electrical conductivity.
Catalog No. 50-276-7014 Supplier Grainger Supplier No. 16U505
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/16" bearing bars at 1-3/16" centers. The panels measure 4' x 8' and have a load capacity of 100 psf. This grating is designed for use in corrosive environments and provides an alternative to traditional metallic gratings. The molded grating has low thermal and electrical conductivity.