Learn More
Grainger Molded Grating Span 3 ft PK2
Catalog No. 502767665
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a fiberglass reinforced plastic grating made of continuous rovings and mat that are molded into panels. It has a high strength-to-weight ratio, corrosion resistance, and is electrically and thermally non-conductive. This grating has a resin and fiberglass construction with a gray color. It has a standard mesh size of 1" x 4" and comes in a variety of panel sizes such as 2' x 8'. The maximum load capacity is 100 psf with a deflection of 0.19" at 30 psf. This grating is designed for use in corrosive environments.
Catalog No. 50-276-7665 Supplier Grainger Supplier No. 16U488
The Fibergrate Fiberglass Molded Grating is a fiberglass reinforced plastic grating made of continuous rovings and mat that are molded into panels. It has a high strength-to-weight ratio, corrosion resistance, and is electrically and thermally non-conductive. This grating has a resin and fiberglass construction with a gray color. It has a standard mesh size of 1" x 4" and comes in a variety of panel sizes such as 2' x 8'. The maximum load capacity is 100 psf with a deflection of 0.19" at 30 psf. This grating is designed for use in corrosive environments.