Learn More
Grainger Molded Grating Span 4 ft.
Catalog No. 502764921
Encompass_Preferred
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/16" bearing bars at 1-3/16" centers. The panels measure 4' x 8' and are 1" thick with a load capacity of 100 psf. This grating is designed for use in corrosive environments and provides an alternative to traditional metal gratings. The molded grating panels allow for easy installation and can be fabricated for custom sizing.
Catalog No. 50-276-4921 Supplier Grainger Supplier No. 16U519
The Fibergrate Fiberglass Molded Grating is a corrosion resistant, high strength to weight ratio grating made of a fiberglass reinforced plastic (FRP) composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/16" bearing bars at 1-3/16" centers. The panels measure 4' x 8' and are 1" thick with a load capacity of 100 psf. This grating is designed for use in corrosive environments and provides an alternative to traditional metal gratings. The molded grating panels allow for easy installation and can be fabricated for custom sizing.