Learn More
Grainger Molded Grating Span 4 ft.
Catalog No. 502766035
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of a fiberglass reinforced plastic composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/8" bearing bars at 1-3/16" centers. The panels are available in a variety of standard sizes up to 4' x 12' and have a load capacity of 100 psf with a maximum deflection of 1/4" at full load. The grating weighs approximately 3 lbs/ft2 and meets OSHA and ADA requirements for slip resistance. With its lightweight, high strength and corrosion resistance, this grating is suitable for industrial platforms, walkways, stairs and more.
Catalog No. 50-276-6035 Supplier Grainger Supplier No. 16U498
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of a fiberglass reinforced plastic composite. It has a matte gray finish and features a square mesh pattern with 1-1/2" x 3/8" bearing bars at 1-3/16" centers. The panels are available in a variety of standard sizes up to 4' x 12' and have a load capacity of 100 psf with a maximum deflection of 1/4" at full load. The grating weighs approximately 3 lbs/ft2 and meets OSHA and ADA requirements for slip resistance. With its lightweight, high strength and corrosion resistance, this grating is suitable for industrial platforms, walkways, stairs and more.