Learn More
Grainger Molded Grating Span 4 ft.
Catalog No. 502768583
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of fiberglass reinforced plastic. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a thickness of 1/2". The grating has a maximum load capacity of 100 psf and meets OSHA and ADA requirements for slip resistance. With its lightweight, high strength design, the Fibergrate grating is suitable for industrial platforms, walkways, stairs and more.
Catalog No. 50-276-8583 Supplier Grainger Supplier No. 16U508
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of fiberglass reinforced plastic. It has a matte gray finish and is available in a standard mesh size of 1" x 3/16" with a thickness of 1/2". The grating has a maximum load capacity of 100 psf and meets OSHA and ADA requirements for slip resistance. With its lightweight, high strength design, the Fibergrate grating is suitable for industrial platforms, walkways, stairs and more.