Learn More
Grainger Molded Grating Span 5 ft.
Catalog No. 502765803
Encompass_Preferred
The Tripp Lite 16U517 is a 19-inch open frame rack enclosure cabinet with a depth of 17 inches. It has an adjustable mounting depth from 14.5 to 17 inches and comes with mounting hardware. The steel enclosure has a black powder coat finish and vented sides for airflow. It has a weight capacity of 200 lbs and 4 adjustable mounting rails. This rack enclosure can house EIA-standard 19-inch rack equipment.
Catalog No. 50-276-5803 Supplier Grainger Supplier No. 16U517
The Tripp Lite 16U517 is a 19-inch open frame rack enclosure cabinet with a depth of 17 inches. It has an adjustable mounting depth from 14.5 to 17 inches and comes with mounting hardware. The steel enclosure has a black powder coat finish and vented sides for airflow. It has a weight capacity of 200 lbs and 4 adjustable mounting rails. This rack enclosure can house EIA-standard 19-inch rack equipment.