Learn More
Grainger Molded Grating Span 5 ft.
Catalog No. 502765622
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous glass fiber rovings bonded with thermosetting polyester resin. It has a matte gray finish and comes in standard panel sizes of 4' x 8' and 4' x 10' with a 1-1/2" square mesh and a 1" depth. It supports up to 100 psf with a deflection of no more than 0.25" and meets OSHA slip resistance requirements. Panels can be cut to custom sizes and installed via banding, bolting, or adhesive bonding. The fiberglass construction ensures durability in corrosive environments.
Catalog No. 50-276-5622 Supplier Grainger Supplier No. 16U515
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous glass fiber rovings bonded with thermosetting polyester resin. It has a matte gray finish and comes in standard panel sizes of 4' x 8' and 4' x 10' with a 1-1/2" square mesh and a 1" depth. It supports up to 100 psf with a deflection of no more than 0.25" and meets OSHA slip resistance requirements. Panels can be cut to custom sizes and installed via banding, bolting, or adhesive bonding. The fiberglass construction ensures durability in corrosive environments.