Learn More
Grainger Molded Grating Span 8 ft.
Catalog No. 502765719
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester or vinyl ester resin. It has a standard mesh size of 1" x 3/16" and comes in a 4' x 8' sheet in a gray color. The grating has a load capacity of 100 psf with a maximum deflection of 1/4" and flame spread rating of Class A per ASTM E84. It is designed for use in corrosive environments and has low electrical and thermal conductivity. The grating weighs approximately 6 lbs/sqft and has a slip-resistant surface.
Catalog No. 50-276-5719 Supplier Grainger Supplier No. 16U501
The Fibergrate Fiberglass Pultruded Grating is a high strength, corrosion resistant grating made of continuous rovings of glass fibers bonded with thermosetting polyester or vinyl ester resin. It has a standard mesh size of 1" x 3/16" and comes in a 4' x 8' sheet in a gray color. The grating has a load capacity of 100 psf with a maximum deflection of 1/4" and flame spread rating of Class A per ASTM E84. It is designed for use in corrosive environments and has low electrical and thermal conductivity. The grating weighs approximately 6 lbs/sqft and has a slip-resistant surface.