Learn More
Grainger Molded Grating Span 8 ft.
Catalog No. 502765808
Encompass_Preferred
The Fibergrate Fiberglass Pultruded Grating is a high-strength, corrosion-resistant fiberglass reinforced plastic (FRP) grating made by a continuous pultrusion process. It has a matte gray finish and is available in a variety of panel sizes and mesh configurations. The grating has a load capacity of 100 psf with a maximum deflection of 0.19" at 30 psf. It meets OSHA and ADA requirements for slip resistance. The grating panels can be easily cut, drilled, or molded for custom applications. Fibergrate grating is ideal for use in corrosive environments.
Catalog No. 50-276-5808 Supplier Grainger Supplier No. 16U522
The Fibergrate Fiberglass Pultruded Grating is a high-strength, corrosion-resistant fiberglass reinforced plastic (FRP) grating made by a continuous pultrusion process. It has a matte gray finish and is available in a variety of panel sizes and mesh configurations. The grating has a load capacity of 100 psf with a maximum deflection of 0.19" at 30 psf. It meets OSHA and ADA requirements for slip resistance. The grating panels can be easily cut, drilled, or molded for custom applications. Fibergrate grating is ideal for use in corrosive environments.