Learn More
Moldex™ 7100 Organic Vapor Cartridge
SDP
Catalog No. 19384301 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Organic vapors; 1 pair/bg
Catalog No.
19-384-301 Organic vapors; 1 pair/bg
1 options
This item is not returnable. View return policy
Catalog No. 19-384-301 Supplier Moldex™ Supplier No. 7100
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with 7000/7800/9000 series reusable respirators
Organic vapors; 1 pair/bg
  • Can be used with 7000, 7800 and 9000 series respirators
  • For oil-free paint sprays/pesticides - use 7100 OV cartridges with 8910 N95 Pre-filter and 7020 retainer
  • For oil-based paint sprays/pesticides - use 7100 OV cartridges with 8970 R95 Pre-filter and 7020 retainer
  • 100% PVC-Free™
Compliance
NIOSH approved.
Specifications
Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH
Product Type Cartridge
For Use With (Equipment) Respirator
Quantity 30/Cs.