Learn More
Moldex™ 7000 Series Respirator Head Strap Assembly
SDP
Catalog No. 19384307 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Head strap assembly
Catalog No.
19-384-307 Head strap assembly
1 options
Catalog No. 19-384-307 Supplier Moldex™ Supplier No. 0072
Add to Cart
Add to Cart
Head strap assembly for use with the 7000 Series half mask respirator.
  • Replacement part
  • Head Strap Assembly for 7000/7800 Series Half Masks Respirators
  • Sold in each quantity or 20/Cs.
Specifications
Disposable Reusable
Product Type Head Harness Assembly
For Use With (Equipment) 7000 series half mask respirator
Includes Head strap and buckles
Model 0072