Learn More
Moldex™ 7200 Acid Gas Cartridge
SDP
Catalog No. 19384302 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Acid gas vapors; 1 pair/bg
Catalog No.
19-384-302 Acid gas vapors; 1 pair/bg
1 options
This item is not returnable. View return policy
Catalog No. 19-384-302 Supplier Moldex™ Supplier No. 7200
This item is not returnable. View return policy
Acid gas cartridges are NIOSH-approved and can be used with 7000, 7800 and 9000 series respirators.
  • NIOSH-approved cartridges protect against the following hazards:
    • Chlorine
    • Hydrogen Chloride
    • Sulfur Dioxide
    • Chlorine Dioxide
    • Hydrogen Fluoride
    • Hydrogen Sulfide
  • Keyless bayonet-style attachment makes assembly quick and easy
  • Pair with 7940 P100, 8970 R95 or 8910 N95 particulate filters
  • For use against particulates when no oil is present use with 8910 N95 Pre-Filter and 7020 retainer
  • For use against particulates when oil is present use with 8970 R95 Pre-Filter and 7020 retainer
  • For contaminants that fall under an OSHA substance specific standard requiring a High Efficiency Particulate Air (HEPA) filter use with 7940 P100 Filter Disk and 7920 Piggyback Adapter
Compliance
NIOSH approved.
Specifications
Cartridge Type Acid Gas
Connection Type Bayonet
Product Type Cartridge
For Use With (Equipment) 7000, 7800 and 9000 series respirators
Quantity 1 Pair