Learn More
Moldex™ 7300 Organic Vapor/Acid Gas Cartridge
SDP
Catalog No. 19384303 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Organic Vapor/Acid Gas; 1 pair/bg
Catalog No.
19-384-303 Organic Vapor/Acid Gas; 1 pair/bg
1 options
This item is not returnable. View return policy
Catalog No. 19-384-303 Supplier Moldex™ Supplier No. 7300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Organic vapor/acid gas cartridges are NIOSH-approved and can be used with 7000, 7800 and 9000 series respirators.
Organic Vapor/Acid Gas; 1 pair/bg
  • Organic vapor/acid gas cartridges are NIOSH-approved for the following hazards:
    • Organic Vapors
    • Chlorine
    • Hydrogen Chloride
    • Sulfur Dioxide or Chlorine Dioxide
    • Hydrogen Sulfide
    • Hydrogen Fluoride
  • Can be used with 7000, 7800 and 9000 series respirators
  • Keyless bayonet-style attachment makes assembly quick and easy
  • Pair with 7940 P100, 8970 R95 or 8910 N95 particulate filters
  • For use against particulates when no oil is present use with 8910 N95 Pre-Filter and 7020 retainer
  • For use against particulates when oil is present use with 8970 R95 Pre-Filter and 7020 retainer
  • For contaminants that fall under an OSHA substance specific standard requiring a High Efficiency Particulate Air (HEPA) filter use with 7940 P100 Filter Disk and 7920 Piggyback Adapter
Compliance
NIOSH approved.
Specifications
Cartridge Type Organic Vapors, Acid Gas
Product Type Cartridge
For Use With (Equipment) Filter
Quantity 30/Cs.