Learn More
Moldex™ 7400 Ammonia/Methylamine Cartridge
SDP
Catalog No. 19384304 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Quantity:
1 Pair
Catalog No. Quantity
19-384-304 1 Pair
1 options
This item is not returnable. View return policy
Catalog No. 19-384-304 Supplier Moldex™ Supplier No. 7400
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with 7000/7800/9000 series reusable respirators
  • Can be used with:
    • Gas proof goggles or Moldex™ 9000 face mask
    • 8910 N95 or 8940 P100 filters against particulates
  • 100% PVC-Free™
Compliance
NIOSH approved.
Specifications
Cartridge Type Ammonia, Methylamine Cartridge
Certifications/Compliance NIOSH
Product Type Cartridge
Quantity 1 Pair