Learn More
Moldex™ 7600 Multi-gas/Vapor Smart Cartridge
SDP
Catalog No. 19384306 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Cartridge Type:
Multi-gas, Vapor Cartridge
Catalog No. Cartridge Type
19-384-306 Multi-gas, Vapor Cartridge
1 options
Catalog No. 19-384-306 Supplier Moldex™ Supplier No. 7600
Add to Cart
Add to Cart
For use with 7000/7800/9000 series reusable respirators
  • Multi-gas/vapor Smart™ cartridge simplifies inventory
  • Approved for nine common gas/vapors
  • Cartridge selection made simpler and misuse reduced
  • Replaces up to five gas/vapor cartridges
  • For use against multi hazards in the same workplace
  • Use with 7600 Multi-gas/vapor Smart™ Assembled Respirators
  • 100% PVC-Free™
Compliance
NIOSH approved.
Specifications
Cartridge Type Multi-gas, Vapor Cartridge
Certifications/Compliance NIOSH
Product Type Multi-Gas/Vapor Cartridge
Quantity 1 Pair