Learn More
Moldex™ 8000 Series Half-Mask Facepieces
SDP
Catalog No. 189991031 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
18-999-1031 Large
18-999-1029 Small
18-999-1030 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-999-1031 Supplier Moldex™ Supplier No. 8003
This item is not returnable. View return policy
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
  • Extra wide surface seals easily and stays sealed even when talking
  • Low profile design provides a large field of vision
  • Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
  • Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifications
Certifications/Compliance NIOSH
Connection Type Snap-in gasket
Material Thermoplastic Elastomer
Product Type Half Mask Facepiece
Format Large