Learn More
Moldex™ 0083 Replacement Exhale Valve Assembly
SDP
Catalog No. 19013473 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Exhale valve assembly
Catalog No.
19-013-473 Exhale valve assembly
1 options
Catalog No. 19-013-473 Supplier Moldex™ Supplier No. 0083
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Replacement exhale valve is for use with 8000 series half mask respirators.
  • Includes holder and diaphragm
  • 100% PVC-Free™
Specifications
Product Type Exhalation Valve
For Use With (Equipment) 8000 Series facepiece
Includes Holder and diaphragm valve
Model 0083