Learn More
Moldex™ Camo Plugs™ Foam Ear Plugs
SDP
Catalog No. 19046811 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded
19-046-811 Corded
19-046-810 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-046-811 Supplier Moldex™ Supplier No. 6609
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unique camouflage-colored foam ear plugs suitable for use around weapons, aircraft, vehicles, watercraft, and other noisy environments. Made of extra-soft, low-pressure foam with a smooth surface for comfort and fit.
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Shaped to fit even very small ear canals
  • Non-irritating smooth surface
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
Cord Color Camouflage
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Industrial, Aircraft, Construction, Watercraft and Environments with Weapons
High Visibility No
Packaging Type Poly bag
Pairs per Package 100 Pairs/Pk.
Plug Material Foam
Plug Color Camouflage
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Product Type Disposable Earplugs
Disposable Single-use
Metal Detectable No