Learn More
MSC Moldex Disposable Particulate Respirator, N95, Universal Size, Flame Resistant, 10/Pk.
Catalog No. 5000677626
Encompass
This disposable particulate respirator provides N95 protection, suitable for humid and hot work conditions. It incorporates a HandyStrap and an exhalation valve to enhance user comfort and ensure compliance with critical safety standards.
  • N95 NIOSH classification
  • Exhalation valve
  • HandyStrap for secure fit
  • Universal size
  • Flame resistant
  • Contains latex components
  • Complies with NIOSH 42 CFR Part 84 and ANSI/ISEA 110-2009 Section 7.11.1
Catalog No. 50-006-77626 Supplier MSC Supplier No. 09679911
This disposable particulate respirator provides N95 protection, suitable for humid and hot work conditions. It incorporates a HandyStrap and an exhalation valve to enhance user comfort and ensure compliance with critical safety standards.
  • N95 NIOSH classification
  • Exhalation valve
  • HandyStrap for secure fit
  • Universal size
  • Flame resistant
  • Contains latex components
  • Complies with NIOSH 42 CFR Part 84 and ANSI/ISEA 110-2009 Section 7.11.1