Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 8940 P100 Filter
SDP

Catalog No. 19003345 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
P100 Particulate Filter/Prefilter Disc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-003-345 P100 Particulate Filter/Prefilter Disc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-345 Supplier Moldex™ Supplier No. 8940

For use with 8000 series half mask respirators

  • P100 Particulate Filter Disk is lighter than traditional Filter Cartridge
  • Piggyback P100 with Moldex™ 8000 gas/vapor cartridges or use standalone filter
  • P100 and R95 can be used for both oil and non-oil based particulates
  • Use P100 Particulate Filter as a prefilter or as a standalone particulate filter
  • Wider field of vision because of low profile
  • Fiberglass-free filters
  • No plastic casing to throw away
  • 100% PVC-Free™

Specifications

Certifications/Compliance NIOSH
Connection Type Threaded
Color Magenta
Filter Type P100
Product Type Particulate Filter
Filter Efficiency 99.97%
For Use With (Application) For protection against both oil and non-oil based particulate aerosols
Quantity 60 Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.