Learn More
Moldex™ Mellows™ Disposable Ear Plugs
SDP
Catalog No. 19384342 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded
19-384-342 Corded
19-384-341 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-384-342 Supplier Moldex™ Supplier No. 6840
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mellows™ disposable ear plugs are made of extra-soft, extra-light foam and are appropriate for mid- to high-level noise areas.
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Non-irritating smooth surface
  • High visibility color makes compliance checks quick and easy
  • Shaped to fit even very small ear canals
Specifications
ANSI S3.1974 Yes
Cord Color Green
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 100 Pairs/Pk.
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 30 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No