Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ M2 Multi-Position Ear Muffs
SDP

Catalog No. 19003359 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Color:
Iridescent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-003-359 Iridescent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-359 Supplier Moldex™ Supplier No. 6200

missing translation for 'validMainframeMsg'

With a lightweight headband and special molded grips, M2 Multi-Position Ear Muffs have adjustability in three different positions including over-the-head, behind-the-neck or under-the-chin.

  • Choose from three position preferences depending on application and comfortability
  • Molded grips make fitting and alignment easy and quick
  • Cup adjustment is highly variable and distributes pressure evenly
  • Available in Black or Special Ops Camouflage color
  • Lightweight plastic headband

Specifications

Color Iridescent
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class A
Cushion Color Black
For Use With (Equipment) Moldex™ Hygiene Kit
Headband Material Plastic
For Use With (Application) Construction, Energy and Utilities, General Manufacturing, Mining, Oil and Gas, Transportation
Model 6200
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB (Over-the-head), 24 dB (Behind-the-head/Under-the-chin)
Product Type M2 Series
Headband Color Black
Dielectric No
Material Plastic
Style Multi-position
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.