Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 8600 Multi-Gas/Vapor Smart™ Cartridges
SDP

Catalog No. 19003344 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Brown shell
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-003-344 Brown shell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-344 Supplier Moldex™ Supplier No. 8600

For use with 8000 series half mask respirators

  • Multi-gas/vapor Smart™ cartridge simplifies inventory
  • Approved for ten common gas/vapors
  • Cartridge selection made simpler and misuse reduced
  • Replaces up to five gas/vapor cartridges
  • For use against multi hazards in the same workplace
  • Use with 8910 N95 or 8940 P100 filters against particulates
  • 100% PVC-Free™

Specifications

Cartridge Type Organic Vapors, Multigas
Certifications (NIOSH) NIOSH 42 CFR 84, TC-23C-1809
Color Brown
Product Type Cartridge
For Use With (Equipment) 8000 series respirator
Quantity 60/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.