Learn More
Moldex™ N95 AirWave™ Respirator with Nuisance Organic Vapor Relief
SDP
Catalog No. 19384350 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Filter Type:
N95
Catalog No. Filter Type
19-384-350 N95
1 options
Catalog No. 19-384-350 Supplier Moldex™ Supplier No. 4800
Add to Cart
Add to Cart
Easy-breathing wave design respirator features an added carbon layer to help filter out ozone and organic vapor odors (less than OSHA PEL).
  • Used against most welding fumes
  • Added carbon layer filters out ozone and organic vapors
  • Soft foam full flange provides comfort
  • Adjustable cool cloth SmartStrap™ for custom fit
  • SoftSpun™ lining
  • Dura-Mesh™ shell for bi-directional protection
Welding, Arc Cutting, Foundry Operations, Brazing, Plasma Spraying, Laser Beam Cutting/Welding, Laboratory Settings, Agriculture, Petrochemical Manufacturing, Rendering Plants, Auto Repair, Waste Disposal
Specifications
Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Dura-Mesh
Product Type Particulate Respirator with Nuisance Organic Vapor Relief
Size Medium/Large
With Exhalation Valve No
Filter Efficiency 95%
Model 4800
Nosepiece Type Molded nose bridge
Nuisance Odor Relief Organic Vapors
Strap Attachment Type SmartStrap™
FDA Cleared Surgical Mask No