Learn More
Description
- P100 particulate filter pre-assembled respirator with Series 7000 facepiece
- Series sleek design for modern, high-tech look and increased worker acceptance
- Lightweight design plus extra-wide sealing area provides all-day comfort
- Low profile, compact design fits well under welding helmets and with safety glasses
- Mask drops down for convenient storage around the neck or locks down for custom fit
- Adjustable head cradle and curved neck buckles for extra comfort
- Wide field of vision
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Connection Type | Bayonet |
| Color | Blue |
| Filter Type | P100 |
| Material | Thermoplastic Elastomer |
| Product Type | Half Mask Respirator |
| Size | Medium |
| For Use With (Application) | Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, pharmaceuticals, petrochemicals |
| For Use With (Equipment) | Series 8910 N95, Series 8970 R95 or Series 7940 P100 filters |
| Includes | P100 Particulate Filter Disks |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
