Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 7000 Series Paint Spray/Pesticide Pre-Assembled Respirators
SDP

Catalog No. 19384326 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-384-326 Small
19-384-327 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-384-326 Supplier Moldex™ Supplier No. 7111
Add to Cart
Add to Cart

Lightweight 7000 Series half-mask respirators are specifically designed to comfortably conform to facial contours and with a modern look. Pre-assembled for oil-free paint spray and pesiticide applications.

  • Paint spray/pesticide pre-assembled respirator with Series 7000 facepiece
  • Includes 7100 organic vapor cartridges, 8910 N95 pre-filters and 7020 retainers
  • Series sleek design for modern, high-tech look and increased worker acceptance
  • Lightweight design plus extra-wide sealing area provides all-day comfort
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Mask drops down for convenient storage around the neck or locks down for custom fit
  • Adjustable head cradle and curved neck buckles for extra comfort
  • Wide field of vision

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Filter Type N95
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Small
For Use With (Application) Pesticide, manufacturing, painting, treatment of timber and wood products, manufacture and use of inks, printing, pharmaceuticals
For Use With (Equipment) Series 8910 N95, Series 8970 R95 or Series 7940 P100 filters
Includes 7000 series facepiece, 7100 organic vapor cartridge, 8910 N95 prefilter and 7020 retainer
Model 7111
Strap Attachment Type Drop-down, Lock-down
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.