Learn More
Moldex™ Pesticide kit
SDP
Catalog No. 19003250C Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Capacity (Metric):
Large
Catalog No. Capacity (Metric)
19-003-250C Large
1 options
Catalog No. 19-003-250C Supplier Moldex™ Supplier No. 8113N
Includes (1) 8000 series facepiece assembly, (5) 8910 N95 pre-filters, (1) 8020 filter retainer and (1 pair) SparkPlugs
Specifications
For Use With (Application) Paint Spray and Pesticide Applications which are Oil-free
For Use With (Equipment) Moldex™ 8000 Series Respirator
Includes One 8000 Facepiece, one pair of 8100 Organic Vapors Cartridges, 8910 N95 Prefilters, one pair of 8020 Retainers, one pair of 6604 SparkPlugs™ foam Earplugs, Paint Bucket
Capacity (Metric) Large