Learn More
Moldex™ SparkPlugs™ PlugStation™ Ear Plug Dispenser
SDP
Catalog No. 19384352 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Metal Detectable:
No
Yes
Holds:
150 Pairs
250 Pairs
500 Pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Metal Detectable Holds
19-384-352 Corded No 150 Pairs
19-384-353 Corded Yes 150 Pairs
19-003-410A Uncorded No 250 Pairs
19-003-410B Uncorded No 500 Pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-384-352 Supplier Moldex™ Supplier No. 6880
Add to Cart
Add to Cart
SparkPlugs™ PlugStation™ dispensers are pre-filled with ear plugs and are convenient for any work environment. Easy access, wall-mountable unit will help ensure compliance. When the PlugStation is empty, simply dispose of it and hang a new one.
  • Holds SparkPlugs™ disposable ear plugs
  • PlugStations™ are sealed to ensure ear plugs stay clean and sanitary
  • High visibility to increase compliance
  • Dispenses one pair at a time to eliminate wasteful grabbing
  • Includes mounting bracket
Specifications
ANSI S3.1974 Yes
Cord Color Purple
Cord Material PVC-free
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
High Visibility Yes
Holds 150 Pairs
Packaging Type Dispenser
Pairs per Package 150 Pairs/Dispenser
Plug Material Foam
Plug Color Multi-color
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Material PVC-free Plastic
Mounting Wall Mountable
Includes Mounting Hardware