Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 8000 Series Half-Mask Pre-Assembled Respirators, Organic Vapor Cartridges
SDP

Catalog No. 18095C Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Organic Vapor; Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-095C Organic Vapor; Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-095C Supplier Moldex™ Supplier No. 8103

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Half-mask respirators conform to the contours of the face and feature an extra-wide seal that remains secure even while talking. Pre-assembled with 8100 Organic Vapor cartridges.

  • Series 8000 facepiece pre-assembled with organic vapor cartridges
  • Comfortable face seal remains sealed even when talking
  • Low-profile design ensures clear vision
  • Strap adjustment points on suspension system allow easy adjustment of straps from cradle
  • Easy and comfortable to wear with safety eye protection
  • Cartridges include inhalation valve and features snap-in mount to eliminate problems caused by misthreading
  • Large valve size promotes easy breathing
  • Baffles on cartridge cap protect filters from sharp objects, dirt and sparks

1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84, TC-23C-1223
Color Gray
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Large
For Use With (Application) Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing
Includes 8000 Facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers
Intrinsic Safety (IS) Rating 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers.
Model 8103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.