Learn More
Moldex™ Pura-Fit™ Disposable Ear Plugs
SDP
Catalog No. p-91437 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Packaging Type:
Paper pouch
Poly bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Packaging Type
17-385 Corded Poly bag
17-384 Uncorded Paper pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-385 Supplier Moldex™ Supplier No. 6900
Only null left
Add to Cart
Edge
Add to Cart
Pura-Fit™ ear plugs are tapered and have a longer shape for easier insertion and grasping . Special foam is formulated for softness and comfort.
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Non-irritating smooth surface
  • Longer length for easy fitting and removal
Specifications
ANSI S3.1974 Yes
Cord Color Green
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 100 Pairs/Pk.
Plug Material Foam
Plug Color Green
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No